Merck
  • Effect of phospholipid composition on an amphipathic peptide-mediated pore formation in bilayer vesicles.
Effect of phospholipid composition on an amphipathic peptide-mediated pore formation in bilayer vesicles.
Biophysical journal (2000-02-02)
F Nicol, S Nir, F C Szoka
ABSTRACT
To better understand the influence of phospholipid acyl-chain composition on the formation of pores by cytotoxic amphipathic helices in biological membranes, the leakage of aqueous contents induced by the synthetic peptide GALA (WEAALAEALAE ALAEHLAEALAEALEALAA) from large unilamellar phospholipid vesicles of various compositions has been studied. Peptide-mediated leakage was examined at pH 5.0 from vesicles made of phosphatidylcholine (PC) and phosphatidylglycerol (PG) with the following acyl-chain compositions: 1-palmitoyl-2-oleoyl (PO), 1,2-dioleoyl (DO), 1, 2-dielaidoyl (DE), and 1,2-dipetroselinoyl (DPe). A mathematical model predicts and simulates the final extents of GALA-mediated leakage of 1-aminonaphthalene-3,6,8-trisulfonic acid (ANTS) and p-xylene-bis-pyridinium bromide (DPX) from 1-palmitoyl-2-oleoyl-phosphatidylcholine/1-palmitoyl-2-oleoyl-phospha tidylglycerol (POPC/POPG) and 1, 2-dielaidoyl-sn-glycero-3-phosphocholine/1, 2-dielaidoyl-phosphatidylglycerol (DEPC/DEPG) liposomes at pH 5.0 as a function of peptide concentration in the bilayer, by considering that GALA pores responsible for this leakage have a minimum size of 10 +/- 2 monomers and are formed by quasiirreversible aggregation of the peptide. With the phospholipid acyl-chain compositions tested, GALA-induced ANTS/DPX leakage follows the rank order POPC/POPG approximately DEPC/DEPG > DPePC/DPePG > DOPC/DOPG. Results from binding experiments reveal that this reduced leakage from DOPC/DOPG vesicles cannot be explained by a reduced binding affinity of the peptide to these membranes. As shown by monitoring the leakage of a fluorescent dextran, an increase in the minimum pore size also does not explain the reduction in ANTS/DPX leakage. The data suggest that surface-associated GALA monomers or aggregates are stabilized in bilayers composed of phospholipids containing a cis unsaturation per acyl chain (DO and DPe), while transbilayer peptide insertion is reduced. GALA-induced ANTS/DPX leakage is also decreased when the vesicles contain phosphatidylethanolamine (PE). This lends further support to the suggestion that factors stabilizing the surface state of the peptide reduce its insertion and subsequent pore formation in the bilayer.
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)