Merck
  • Epoxomicin, a potent and selective proteasome inhibitor, exhibits in vivo antiinflammatory activity.
Epoxomicin, a potent and selective proteasome inhibitor, exhibits in vivo antiinflammatory activity.
Proceedings of the National Academy of Sciences of the United States of America (1999-09-01)
L Meng, R Mohan, B H Kwok, M Elofsson, N Sin, C M Crews
ABSTRACT
The proteasome regulates cellular processes as diverse as cell cycle progression and NF-kappaB activation. In this study, we show that the potent antitumor natural product epoxomicin specifically targets the proteasome. Utilizing biotinylated-epoxomicin as a molecular probe, we demonstrate that epoxomicin covalently binds to the LMP7, X, MECL1, and Z catalytic subunits of the proteasome. Enzymatic analyses with purified bovine erythrocyte proteasome reveal that epoxomicin potently inhibits primarily the chymotrypsin-like activity. The trypsin-like and peptidyl-glutamyl peptide hydrolyzing catalytic activities also are inhibited at 100- and 1,000-fold slower rates, respectively. In contrast to peptide aldehyde proteasome inhibitors, epoxomicin does not inhibit nonproteasomal proteases such trypsin, chymotrypsin, papain, calpain, and cathepsin B at concentrations of up to 50 microM. In addition, epoxomicin is a more potent inhibitor of the chymotrypsin-like activity than lactacystin and the peptide vinyl sulfone NLVS. Epoxomicin also effectively inhibits NF-kappaB activation in vitro and potently blocks in vivo inflammation in the murine ear edema assay. These results thus define epoxomicin as a novel proteasome inhibitor that likely will prove useful in exploring the role of the proteasome in various in vivo and in vitro systems.
Sigma-Aldrich
Epoxomicin, ≥95% (HPLC), solid
Pricing and availability is not currently available.