Merck
  • Isotope-labeling of the fibril binding compound FSB via a Pd-catalyzed double alkoxycarbonylation.
Isotope-labeling of the fibril binding compound FSB via a Pd-catalyzed double alkoxycarbonylation.
The Journal of organic chemistry (2012-05-23)
Mia N Burhardt, Rolf Taaning, Niels Chr Nielsen, Troels Skrydstrup
ABSTRACT
We have synthesized two isotopically labeled variants of the β-amyloid binding compound FSB possessing (13)C-labels on the two terminal aryl carboxylic acid moieties. One of these was also fully deuterated on the olefinic spacers. The (13)C-isotope labeling was achieved applying a Pd-catalyzed methoxycarbonylation of the corresponding aryl chlorides with externally (ex situ) generated (13)C-labeled CO. Application of the Shirakawa-Hayashi protocol for the Pd-catalyzed reduction of a dialkyne intermediate using D(2)O allowed for the selective deuterium labeling of the two trans-C,C double bonds of FSB.
Sigma-Aldrich
9-Methyl-9H-fluorene-9-carbonyl-13C chloride, ≥97.0% (GC)