Merck
  • Purification and characterization of maltooligosaccharide-forming α-amylase from moderately halophilic Marinobacter sp. EMB8.
Purification and characterization of maltooligosaccharide-forming α-amylase from moderately halophilic Marinobacter sp. EMB8.
Bioresource technology (2011-12-27)
Sumit Kumar, S K Khare
ABSTRACT
Maltooligosaccharides especially maltotriose and maltotetraose producing amylases are highly desirable for application in bread making and other food industries. A maltotriose and maltotetraose producing amylase from moderately halophilic Marinobacter sp. EMB8 is described. Under optimized culture conditions, 48.0 IU/mL amylase was obtained. The enzyme was purified to homogeneity by ultrafiltration, DEAE cellulose and Sephadex G-75 column chromatography with 52% yield and 76-fold purification. It was a monomeric protein of 72 kDa. The amylase had many novel features viz. stability up to 20% NaCl, 80 °C temperature, pH 6.0-11.0 and in wide range of organic solvents at high concentrations. The enzyme efficiently hydrolyzed starch into maltooligosaccharides rich in maltotriose and maltotetraose. These novel properties make the Marinobacter sp. amylase a potentially useful enzyme.
Sigma-Aldrich
Maltotetraose, 94% (HPLC)