Merck
The CXC-chemokine, H174: expression in the central nervous system.
Journal of neurovirology (1999-03-05)
Y Luo, R Kim, D Gabuzda, S Mi, L A Collins-Racie, Z Lu, K A Jacobs, M E Dorf
ABSTRACT
H174 is a new member of the CXC-chemokine family. A cDNA probe containing the entire H174 coding region recognized a predominant inducible transcript of approximately 1.5 kb expressed in interferon (IFN) activated astrocytoma and monocytic cell lines. H174 message can be induced following IFN-alpha, IFN-beta, or IFN-gamma stimulation. H174 message was also detected in IFN treated cultures of primary human astrocytes, but was absent in unstimulated astrocytes. H174, like IP10 and Mig, lacks the ELR sequence associated with the neutrophil specificity characteristic of most CXC-chemokines. Preliminary experiments suggest H174, IP10 and Mig are independently regulated. Recombinant H174 is a weak chemoattractant for monocyte-like cells. H174 can also stimulate calcium flux responses. The data support the classification of H174 as a member of a subfamily of interferon-gamma inducible non-ELR CXC-chemokines. Brain tissues were obtained at autopsy from one patient with AIDS dementia, one patient with multiple sclerosis, and two normal control patients. H174 and Mig were detected by RT-PCR in brain tissue cDNA derived from the patients with pathological conditions associated with activated astrocytes but not in cDNA from control specimens.
Sigma-Aldrich
Interferon-Inducible T Cell α Chemoattractant human, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture