Merck
  • Promoter analyses and transcriptional profiling of eggplant polyphenol oxidase 1 gene (SmePPO1) reveal differential response to exogenous methyl jasmonate and salicylic acid.
Promoter analyses and transcriptional profiling of eggplant polyphenol oxidase 1 gene (SmePPO1) reveal differential response to exogenous methyl jasmonate and salicylic acid.
Journal of plant physiology (2012-03-02)
Santoshkumar M Shetty, Arun Chandrashekar, Yeldur P Venkatesh
ABSTRACT
The transcriptional regulation of multigenic eggplant (Solanum melongena) polyphenol oxidase genes (SmePPO) is orchestrated by their corresponding promoters which mediate developmentally regulated expression in response to myriad biotic and abiotic factors. However, information on structural features of SmePPO promoters and modulation of their expression by plant defense signals are lacking. In the present study, SmePPOPROMOTERs were cloned by genome walking, and their transcription start sites (TSS) were determined by RLM-RACE. Extensive sequence analyses revealed the presence of evolutionarily conserved and over-represented putative cis-acting elements involved in light-regulated transcription, biosynthetic pathways (phenylpropanoid/flavonoid), hormone signaling (abscisic acid, gibberellic acid, jasmonate and salicylate), elicitor and stress responses (cold/dehydration responses), sugar metabolism and plant defense signaling (W-BOX/WRKY) that are common to SmePPOPROMOTER1 and 2. The TSS for SmePPO genes are located 9-15bp upstream of ATG with variable lengths of 5' untranslated regions. Transcriptional profiling of SmePPOs in eggplant seedlings has indicated differential response to methyl jasmonate (MeJA) or salicylic acid (SA) treatment. In planta, while MeJA elicited expression of all the six SmePPOs, SA was only able to induce the expression of SmePPO4-6. Interestingly, in dual treatment, SA considerably repressed the MeJA-induced expression of SmePPOs. Functional dissection of SmePPOPROMOTER1 by deletion analyses using Agrobacterium-mediated transient expression in tobacco leaves has shown that MeJA enhances the SmePPOPROMOTER1-β-glucuronidase (GUS) expression in vivo, while SA does not. Histochemical and quantitative GUS assays have also indicated the negative effect of SA on MeJA-induced expression of SmePPOPROMOTER1. By combining in silico analyses, transcriptional profiling and expression of SmePPOPROMOTER1-GUS fusions, the role of SA on the modulation of MeJA-induced SmePPO1 expression has been elucidated. It is concluded that similar to the coding regions of multigenic SmePPOs, the regulatory elements are also evolutionarily conserved and fall into two distinct sub-classes based on their responses to MeJA and SA.