Merck
  • Rapid method for the detection of genetically engineered microorganisms by polymerase chain reaction from soil and sediments.
Rapid method for the detection of genetically engineered microorganisms by polymerase chain reaction from soil and sediments.
Journal of industrial microbiology & biotechnology (1998-06-27)
A A Khan, R A Jones, C E Cerniglia
ABSTRACT
A rapid and sensitive method for the detection of genetically engineered microorganisms in soil and sediments has been devised by in vitro amplification of the target DNAs by a polymerase chain reaction. A cloned catechol 2,3-dioxygenase gene located on the recombinant plasmid pOH101 was transferred to Pseudomonas putida MMB2442 by triparental crossing and used as a target organism. For the polymerase chain reaction from soil and sediment samples, the template DNA was released from a 100-mg soil sample. Bacterial seeded soil samples were washed with Tris-EDTA buffer (pH 8.0) and treated with a detergent lysis solution at 100 degrees C. After addition of 1% polyvinylpolypyrrolidine solution, the samples were boiled for 5 min. Supernatant containing nucleic acid was purified with a PCR purification kit. The purified DNA was subjected to polymerase chain reaction, using two specific primers designed for the amplification of catechol 2,3-dioxygenase gene sequences. The detection limit was 10(2) cells per gram of soil. This method is rapid and obviates the need for lengthy DNA purification from soil samples.