Merck
  • Vasoinhibin is Generated and Promotes Inflammation in Mild Antigen-induced Arthritis.
Vasoinhibin is Generated and Promotes Inflammation in Mild Antigen-induced Arthritis.
Endocrinology (2022-03-20)
Georgina Ortiz, Maria G Ledesma-Colunga, Zhijian Wu, Jose F García-Rodrigo, Norma Adan, Oscar F Martinez-Diaz, Ericka A De Los Ríos, Fernando López-Barrera, Gonzalo Martínez de la Escalera, Carmen Clapp
ABSTRACT
Inflammatory arthritis defines a family of diseases influenced by reproductive hormones. Vasoinhibin, a fragment of the hormone prolactin (PRL), has antiangiogenic and proinflammatory properties. We recently showed that vasoinhibin reduces joint inflammation and bone loss in severe antigen-induced arthritis (AIA) by an indirect mechanism involving the inhibition of pannus vascularization. This unexpected finding led us to hypothesize that a severe level of inflammation in AIA obscured the direct proinflammatory action of vasoinhibin while allowing the indirect anti-inflammatory effect via its antiangiogenic properties. In agreement with this hypothesis, here we show that the intra-articular injection of an adeno-associated virus type-2 vector encoding vasoinhibin reduced joint inflammation in a severe AIA condition, but elevated joint inflammation in a mild AIA model. The proinflammatory effect, unmasked in mild AIA, resulted in joint swelling, enhanced leukocyte infiltration, and upregulation of expression of genes encoding proinflammatory mediators (Il1b, Il6, Inos, Mmp3), adhesion molecule (Icam1), and chemokines (Cxcl1, Cxcl2, Cxcl3, Ccl2). Furthermore, vasoinhibin induced the expression of proinflammatory mediators and chemokines in cultured synovial fibroblasts through nuclear factor-κB. Finally, matrix metalloproteases and cathepsin D, upregulated in the arthritic joint, cleaved PRL to vasoinhibin, and vasoinhibin levels increased in the circulation of mice subjected to AIA. We suggest that vasoinhibin is generated during inflammatory arthritis and acts on synovial fibroblasts and endothelial cells to initially promote and later inhibit inflammation, respectively. These opposite effects may work together to help keep joint inflammation under balance.
Sigma-Aldrich
Anti-iNOS/NOS II Antibody, NT, from rabbit, purified by affinity chromatography