Merck
  • Suspension culture in a rotating bioreactor for efficient generation of human intestinal organoids.
Suspension culture in a rotating bioreactor for efficient generation of human intestinal organoids.
Cell reports methods (2022-12-02)
Junichi Takahashi, Tomohiro Mizutani, Hady Yuki Sugihara, Sayaka Nagata, Shu Kato, Yui Hiraguri, Sayaka Takeoka, Mao Tsuchiya, Reiko Kuno, Sei Kakinuma, Mamoru Watanabe, Ryuichi Okamoto
ABSTRACT
Human intestinal organoids (HIOs) derived from human pluripotent stem cells (hPSCs) hold great promise for translational medical applications. A common method to obtain HIOs has been to harvest floating hindgut spheroids arising from hPSCs. As this technique is elegant but burdensome due to the complex protocol and line-to-line variability, a more feasible method is desired. Here, we establish a robust differentiation method into suspension-cultured HIOs (s-HIOs) by seeding dissociated cells on a spheroid-forming plate. This protocol realizes the reliable generation of size-controllable spheroids. Under optimized conditions in a rotating bioreactor, the generated spheroids quickly grow and mature into large s-HIOs with supporting mesenchyme. Upon mesenteric transplantation, s-HIOs further mature and develop complex tissue architecture in vivo. This method demonstrates that intestinal tissue can be generated from iPSC-derived HIOs via suspension induction and bioreactor maturation, establishing a reliable culture platform with wide applications in regenerative medicine.
Sigma-Aldrich
Anti-LYZ antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution