Merck
  • Choice of template delivery mitigates the genotoxic risk and adverse impact of editing in human hematopoietic stem cells.
Choice of template delivery mitigates the genotoxic risk and adverse impact of editing in human hematopoietic stem cells.
Cell stem cell (2022-10-08)
Samuele Ferrari, Aurelien Jacob, Daniela Cesana, Marianne Laugel, Stefano Beretta, Angelica Varesi, Giulia Unali, Anastasia Conti, Daniele Canarutto, Luisa Albano, Andrea Calabria, Valentina Vavassori, Carlo Cipriani, Maria Carmina Castiello, Simona Esposito, Chiara Brombin, Federica Cugnata, Oumeya Adjali, Eduard Ayuso, Ivan Merelli, Anna Villa, Raffaella Di Micco, Anna Kajaste-Rudnitski, Eugenio Montini, Magalie Penaud-Budloo, Luigi Naldini
ABSTRACT
Long-range gene editing by homology-directed repair (HDR) in hematopoietic stem/progenitor cells (HSPCs) often relies on viral transduction with recombinant adeno-associated viral vector (AAV) for template delivery. Here, we uncover unexpected load and prolonged persistence of AAV genomes and their fragments, which trigger sustained p53-mediated DNA damage response (DDR) upon recruiting the MRE11-RAD50-NBS1 (MRN) complex on the AAV inverted terminal repeats (ITRs). Accrual of viral DNA in cell-cycle-arrested HSPCs led to its frequent integration, predominantly in the form of transcriptionally competent ITRs, at nuclease on- and off-target sites. Optimized delivery of integrase-defective lentiviral vector (IDLV) induced lower DNA load and less persistent DDR, improving clonogenic capacity and editing efficiency in long-term repopulating HSPCs. Because insertions of viral DNA fragments are less frequent with IDLV, its choice for template delivery mitigates the adverse impact and genotoxic burden of HDR editing and should facilitate its clinical translation in HSPC gene therapy.
Sigma-Aldrich
DAPI, for nucleic acid staining