Merck
  • Bundle-sheath cell regulation of xylem-mesophyll water transport via aquaporins under drought stress: a target of xylem-borne ABA?
Bundle-sheath cell regulation of xylem-mesophyll water transport via aquaporins under drought stress: a target of xylem-borne ABA?
The Plant journal : for cell and molecular biology (2011-03-16)
Arava Shatil-Cohen, Ziv Attia, Menachem Moshelion
ABSTRACT
The hydraulic conductivity of the leaf vascular system (K(leaf) ) is dynamic and decreases rapidly under drought stress, possibly in response to the stress phytohormone ABA, which increases sharply in the xylem sap (ABA(xyl) ) during periods of drought. Vascular bundle-sheath cells (BSCs; a layer of parenchymatous cells tightly enwrapping the entire leaf vasculature) have been hypothesized to control K(leaf) via the specific activity of BSC aquaporins (AQPs). We examined this hypothesis and provide evidence for drought-induced ABA(xyl) diminishing BSC osmotic water permeability (P(f) ) via downregulated activity of their AQPs. ABA fed to the leaf via the xylem (petiole) both decreased K(leaf) and led to stomatal closure, replicating the effect of drought. In contrast, smearing ABA on the leaf blade, while also closing stomata, did not decrease K(leaf) within 2-3 h of application, demonstrating that K(leaf) does not depend entirely on stomatal closure. GFP-labeled BSCs showed decreased P(f) in response to 'drought' and ABA treatment, and a reversible decrease with HgCl(2) (an AQP blocker). These P(f) responses, absent in mesophyll cells, suggest stress-regulated AQP activity specific to BSCs, and imply a role for these cells in decreasing K(leaf) via a reduction in P(f) . Our results support the above hypothesis and highlight the BSCs as hitherto overlooked vasculature sensor compartments, extending throughout the leaf and functioning as 'stress-regulated valves' converting vasculature chemical signals (possibly ABA(xyl) ) into leaf hydraulic signals.