Merck
  • Impaired sterol ester synthesis alters the response of Arabidopsis thaliana to Phytophthora infestans.
Impaired sterol ester synthesis alters the response of Arabidopsis thaliana to Phytophthora infestans.
The Plant journal : for cell and molecular biology (2012-10-18)
Michaela Kopischke, Lore Westphal, Korbinian Schneeberger, Richard Clark, Stephan Ossowski, Vera Wewer, René Fuchs, Jörn Landtag, Gerd Hause, Peter Dörmann, Volker Lipka, Detlef Weigel, Paul Schulze-Lefert, Dierk Scheel, Sabine Rosahl
ABSTRACT
Non-host resistance of Arabidopsis thaliana against Phytophthora infestans, the causal agent of late blight disease of potato, depends on efficient extracellular pre- and post-invasive resistance responses. Pre-invasive resistance against P. infestans requires the myrosinase PEN2. To identify additional genes involved in non-host resistance to P. infestans, a genetic screen was performed by re-mutagenesis of pen2 plants. Fourteen independent mutants were isolated that displayed an enhanced response to Phytophthora (erp) phenotype. Upon inoculation with P. infestans, two mutants, pen2-1 erp1-3 and pen2-1 erp1-4, showed an enhanced rate of mesophyll cell death and produced excessive callose deposits in the mesophyll cell layer. ERP1 encodes a phospholipid:sterol acyltransferase (PSAT1) that catalyzes the formation of sterol esters. Consistent with this, the tested T-DNA insertion lines of PSAT1 are phenocopies of erp1 plants. Sterol ester levels are highly reduced in all erp1/psat1 mutants, whereas sterol glycoside levels are increased twofold. Excessive callose deposition occurred independently of PMR4/GSL5 activity, a known pathogen-inducible callose synthase. A similar formation of aberrant callose deposits was triggered by the inoculation of erp1 psat1 plants with powdery mildew. These results suggest a role for sterol conjugates in cell non-autonomous defense responses against invasive filamentous pathogens.