Merck
IL-12 instructs skin homing of human Th2 cells.
Journal of immunology (Baltimore, Md. : 1950) (2006-09-05)
Tilo Biedermann, Günther Lametschwandtner, Kirsten Tangemann, Julia Kund, Sonja Hinteregger, Nicole Carballido-Perrig, Antal Rot, Christoph Schwärzler, José M Carballido
ABSTRACT
Distinct pattern of homing receptors determines the tissue preference for T cells to exert their effector functions. This homing competence is mostly determined early during T cell activation of naive T cells. In contrast, mechanisms governing the acquisition of particular homing receptors by T cells of the memory phenotype remain enigmatic. Th2 cell-mediated allergic diseases tend to flare during infections despite that these infections prime APCs to produce the prototypic Th1 cell-differentiating cytokine IL-12. In this study, we investigate the effect of IL-12 on the regulation of cutaneous lymphocyte Ag (CLA) on differentiated Th2 cells and consequences of this expression for allergic inflammation. Upon activation with IL-12, CLA- Th2 cells rapidly up-regulated IL-12Rbeta2 chain, alpha(1-3)-fucosyltransferase VII, and CLA molecules. IL-12-mediated CLA expression on Th2 cells was functional because it mediated rolling of these Th2 cells on E-selectin in vitro and migration into human skin grafts in SCID mice. CLA induction occurred immediately after exposure to IL-12 and was independent of IFN-gamma expression. In accordance, the transcription factor mediating IFN-gamma expression, T-bet, does not directly affect CLA expression. However, CLA expression was further enhanced after IL-12 treatment of T-bet+ -transfected Th2 cells in agreement with an increased IL-12 responsiveness of these cells caused by T-bet. The finding that IL-12 conferred skin-homing potential to already differentiated Th2 cells before inducing a switch in their cytokine production profile may explain the observed exacerbation of allergic skin diseases following bacterial infections.