Merck
  • Optimization of High-Throughput Multiplexed Phenotyping of Extracellular Vesicles Performed in 96-Well Microtiter Plates.
Optimization of High-Throughput Multiplexed Phenotyping of Extracellular Vesicles Performed in 96-Well Microtiter Plates.
Polymers (2021-07-25)
Malene Møller Jørgensen, Jenni Kathrine Sloth, Rikke Bæk
ABSTRACT
Extracellular vesicles (EVs) are promising biomarkers for several diseases, however, no simple and robust methods exist to characterize EVs in a clinical setting. The EV Array analysis is based on a protein microarray platform, where antibodies are printed onto a solid surface that enables the capture of small EVs (sEVs) by their surface or surface-associated proteins. The EV Array analysis was transferred to an easily handled microtiter plate (MTP) format and a range of optimization experiments were performed within this study. The optimization was performed in a comprehensive analytical setup where the focus was on the selection of additives added to spotting-, blocking-, and incubation buffers as well as the storage of printed antibody arrays under different temperatures from one day to 12 weeks. After ending the analysis, the stability of the fluorescent signal was investigated at different storage conditions for up to eight weeks. The various parameters and conditions tested within this study were shown to have a high influence on each other. The reactivity of the spots was found to be preserved for up to 12 weeks when stored at room temperature and using blocking procedure IV in combination with trehalose in the spotting buffer. Similar preservation could be obtained using glycerol or sciSPOT D1 in the spotting buffers, but only if stored at 4 °C after blocking procedure I. Conclusively, it was found that immediate scanning of the MTPs after analysis was not critical if stored dried, in the dark, and at room temperature. The findings in this study highlight the necessity of performing optimization experiments when transferring an established analysis to a new technological platform.