Merck
  • Requirement of DNMT1 to orchestrate epigenomic reprogramming for NPM-ALK-driven lymphomagenesis.
Requirement of DNMT1 to orchestrate epigenomic reprogramming for NPM-ALK-driven lymphomagenesis.
Life science alliance (2020-12-15)
Elisa Redl, Raheleh Sheibani-Tezerji, Crhistian de Jesus Cardona, Patricia Hamminger, Gerald Timelthaler, Melanie Rosalia Hassler, Maša Zrimšek, Sabine Lagger, Thomas Dillinger, Lorena Hofbauer, Kristina Draganić, Andreas Tiefenbacher, Michael Kothmayer, Charles H Dietz, Bernard H Ramsahoye, Lukas Kenner, Christoph Bock, Christian Seiser, Wilfried Ellmeier, Gabriele Schweikert, Gerda Egger
ABSTRACT
Malignant transformation depends on genetic and epigenetic events that result in a burst of deregulated gene expression and chromatin changes. To dissect the sequence of events in this process, we used a T-cell-specific lymphoma model based on the human oncogenic nucleophosmin-anaplastic lymphoma kinase (NPM-ALK) translocation. We find that transformation of T cells shifts thymic cell populations to an undifferentiated immunophenotype, which occurs only after a period of latency, accompanied by induction of the MYC-NOTCH1 axis and deregulation of key epigenetic enzymes. We discover aberrant DNA methylation patterns, overlapping with regulatory regions, plus a high degree of epigenetic heterogeneity between individual tumors. In addition, ALK-positive tumors show a loss of associated methylation patterns of neighboring CpG sites. Notably, deletion of the maintenance DNA methyltransferase DNMT1 completely abrogates lymphomagenesis in this model, despite oncogenic signaling through NPM-ALK, suggesting that faithful maintenance of tumor-specific methylation through DNMT1 is essential for sustained proliferation and tumorigenesis.
Sigma-Aldrich
Anti-CDK6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution