Merck
  • Galectin-3: role in ocular allergy and potential as a predictive biomarker.
Galectin-3: role in ocular allergy and potential as a predictive biomarker.
The British journal of ophthalmology (2018-03-05)
Frans Eberth Costa Andrade, Mab Pereira Corrêa, Alexandre Dantas Gimenes, Myrna Serapião Dos Santos, Mauro Campos, Roger Chammas, José Álvaro Pereira Gomes, Cristiane Damas Gil
ABSTRACT
To evaluate galectin-3 (Gal-3), a β-galactoside binding protein, as a possible biomarker in ocular allergy and further investigated the role of endogenous Gal-3 in a murine model of ovalbumin (OVA)-induced allergic conjunctivitis (AC). Conjunctival impression cytology specimens from control and patients with severe vernal keratoconjunctivitis, treated or untreated, were used to evaluate Gal-3 expression by immunocytochemistry. To investigate the mechanism of action of Gal-3, OVA-immunised BALB/c male wild-type (WT) and Gal-3 null (Gal-3-/-) mice were challenged with eye drops containing OVA on days 14-16 with a subset of animals pretreated with 0.03% tacrolimus (TC) or dexamethasone (Dex). Patients with AC and OVA-sensitised WT mice exhibited increased levels of Gal-3 in the conjunctiva compared with control, an effect reverted by the action of Dex and TC therapy. Twenty-four hours after the final OVA challenge, total and anti-OVA IgE levels increased significantly in the blood of OVA-sensitised WT and Gal-3-/- mice compared with controls, supporting the efficacy of the AC model. The lack of endogenous Gal-3 exacerbated the local inflammatory response, increasing the influx of eosinophils and mast cell activation. Additionally, OVA-sensitised Gal-3-/- animals exhibited increased CD4+ expression in the eyes as well as eotaxin, IL-4, IL-13 and interferon-γ levels in the tear fluid compared with WT animals. Gal-3 contributes to the pathogenesis of ocular allergy and represents a relevant therapeutic target.
Product Description
Millipore
MILLIPLEX® Mouse TH17 Panel, Premixed Mouse Th17 25-Plex Panel