Merck
  • Changes in subtypes of Ca microdomains following partial injury to the central nervous system.
Changes in subtypes of Ca microdomains following partial injury to the central nervous system.
Metallomics : integrated biometal science (2014-01-16)
Ivan Lozić, Carole A Bartlett, Jeremy A Shaw, K Swaminathan Iyer, Sarah A Dunlop, Matt R Kilburn, Melinda Fitzgerald
ABSTRACT
Rapid changes in Ca(2+) concentration and location in response to injury play key roles in a range of biological systems. However, quantitative analysis of changes in size and distribution of Ca(2+) microdomains in specific cell types in whole tissue samples has been limited by analytical resolution and reliance on indirect Ca(2+) indicator systems. Here, we combine the unique advantages of nanoscale secondary ion mass spectrometry (NanoSIMS) with immunohistochemistry to directly quantify changes in number, size and intensity of Ca microdomains specific to axonal or glial regions vulnerable to spreading damage following neurotrauma. Furthermore, using NanoSIMS allows separate quantification of Ca microdomains according to their co-localization with areas enriched in P. We rapidly excise and cryopreserve optic nerve segments from adult rat at time points ranging from 5 minutes to 3 months after injury, allowing assessment of Ca microdomains dynamics with minimal disruption due to tissue processing. We demonstrate significantly more non-P co-localized Ca microdomains in glial than axonal regions in normal optic nerve. The density of Ca microdomains not co-localized with areas enriched in P rapidly, selectively and significantly decreases after injury; densities of Ca microdomains co-localized with P enriched areas are unchanged. An efflux of Ca(2+) from microdomains not co-localized with P may contribute to the structural and functional deficits observed in nerve vulnerable to spreading damage following neurotrauma. NanoSIMS analyses of Ca microdomains allow quantitative and novel insights into Ca dynamics, applicable to a range of normal, as well as diseased or injured mammalian systems.
Sigma-Aldrich
Anti-VDAC2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution