Merck
  • CDKD-dependent activation of CDKA;1 controls microtubule dynamics and cytokinesis during meiosis.
CDKD-dependent activation of CDKA;1 controls microtubule dynamics and cytokinesis during meiosis.
The Journal of cell biology (2020-07-02)
Kostika Sofroni, Hirotomo Takatsuka, Chao Yang, Nico Dissmeyer, Shinichiro Komaki, Yuki Hamamura, Lev Böttger, Masaaki Umeda, Arp Schnittger
ABSTRACT
Precise control of cytoskeleton dynamics and its tight coordination with chromosomal events are key to cell division. This is exemplified by formation of the spindle and execution of cytokinesis after nuclear division. Here, we reveal that the central cell cycle regulator CYCLIN DEPENDENT KINASE A;1 (CDKA;1), the Arabidopsis homologue of Cdk1 and Cdk2, partially in conjunction with CYCLIN B3;1 (CYCB3;1), is a key regulator of the microtubule cytoskeleton in meiosis. For full CDKA;1 activity, the function of three redundantly acting CDK-activating kinases (CAKs), CDKD;1, CDKD;2, and CDKD;3, is necessary. Progressive loss of these genes in combination with a weak loss-of-function mutant in CDKA;1 allowed a fine-grained dissection of the requirement of cell-cycle kinase activity for meiosis. Notably, a moderate reduction of CDKA;1 activity converts the simultaneous cytokinesis in Arabidopsis, i.e., one cytokinesis separating all four meiotic products concurrently into two successive cytokineses with cell wall formation after the first and second meiotic division, as found in many monocotyledonous species.
Sigma-Aldrich
StrepTag II Monoclonal Antibody, Novagen®