Merck
Polymorphisms and mutations of human TMPRSS6 in iron deficiency anemia.
Blood cells, molecules & diseases (2009-10-13)
E Beutler, C Van Geet, D M W M te Loo, T Gelbart, K Crain, J Truksa, P L Lee
ABSTRACT
Male subjects with iron deficiency from the general population were examined for polymorphisms or sporadic mutations in TMPRSS6 to identify genetic risk factors for iron deficiency anemia. Three uncommon non-synonymous polymorphisms were identified, G228D, R446W, and V795I (allele frequencies 0.0074, 0.023 and 0.0074 respectively), of which the R446W polymorphism appeared to be overrepresented in the anemic population. In addition, three children with iron refractory iron deficiency anemia, and one sibling with iron responsive iron deficiency anemia were also examined for polymorphisms or sporadic mutations in TMPRSS6. Two children (family 1) were compound heterozygotes for a L674F mutation and a previously described splicing defect predicted to cause skipping of exon 13 (IVS13+1 G>A). One child from the second family was homozygous for a deletion (497T) causing a frameshift (L166X+36) and premature termination. The sibling and mother from the second family were compound heterozygotes for the L166X mutation and the uncommon R446W polymorphism. Although in vitro expression studies demonstrated that the R446W isoform was biologically similar to wildtype Tmprss6, clinical data indicate that the R446W produces a milder disease when carried in trans with severe mutation in Tmprss6. The four children carrying mutations in TMPRSS6 all exhibited inappropriately high urinary hepcidin levels for the degree of iron deficiency.