Merck
  • Loss of NAPRT1 expression by tumor-specific promoter methylation provides a novel predictive biomarker for NAMPT inhibitors.
Loss of NAPRT1 expression by tumor-specific promoter methylation provides a novel predictive biomarker for NAMPT inhibitors.
Clinical cancer research : an official journal of the American Association for Cancer Research (2013-10-08)
David S Shames, Kristi Elkins, Kimberly Walter, Thomas Holcomb, Pan Du, Dane Mohl, Yang Xiao, Thinh Pham, Peter M Haverty, Bianca Liederer, Xiaorong Liang, Robert L Yauch, Thomas O'Brien, Richard Bourgon, Hartmut Koeppen, Lisa D Belmont
ABSTRACT
We sought to identify predictive biomarkers for a novel nicotinamide phosphoribosyltransferase (NAMPT) inhibitor. We use a NAMPT inhibitor, GNE-617, to evaluate nicotinic acid rescue status in a panel of more than 400 cancer cell lines. Using correlative analysis and RNA interference (RNAi), we identify a specific biomarker for nicotinic acid rescue status. We next determine the mechanism of regulation of expression of the biomarker. Finally, we develop immunohistochemical (IHC) and DNA methylation assays and evaluate cancer tissue for prevalence of the biomarker across indications. Nicotinate phosphoribosyltransferase (NAPRT1) is necessary for nicotinic acid rescue and its expression is the major determinant of rescue status. We demonstrate that NAPRT1 promoter methylation accounts for NAPRT1 deficiency in cancer cells, and NAPRT1 methylation is predictive of rescue status in cancer cell lines. Bisulfite next-generation sequencing mapping of the NAPRT1 promoter identified tumor-specific sites of NAPRT1 DNA methylation and enabled the development of a quantitative methylation-specific PCR (QMSP) assay suitable for use on archival formalin-fixed paraffin-embedded tumor tissue. Tumor-specific promoter hypermethylation of NAPRT1 inactivates one of two NAD salvage pathways, resulting in synthetic lethality with the coadministration of a NAMPT inhibitor. NAPRT1 expression is lost due to promoter hypermethylation in most cancer types evaluated at frequencies ranging from 5% to 65%. NAPRT1-specific immunohistochemical or DNA methylation assays can be used on archival formalin paraffin-embedded cancer tissue to identify patients likely to benefit from coadministration of a Nampt inhibitor and nicotinic acid.
Product Description
Roche
High Pure FFPE RNA Micro Kit, kit of for 50 isolations