Merck
  • Ganoderic acids suppress growth and invasive behavior of breast cancer cells by modulating AP-1 and NF-kappaB signaling.
Ganoderic acids suppress growth and invasive behavior of breast cancer cells by modulating AP-1 and NF-kappaB signaling.
International journal of molecular medicine (2008-04-22)
Jiahua Jiang, Brian Grieb, Anita Thyagarajan, Daniel Sliva
ABSTRACT
Structurally related lanostane-type triterpenes, ganoderic acid A, F and H (GA-A, GA-F, GA-H), were identified in an oriental medicinal mushroom Ganoderma lucidum. In the present study we evaluated the effect of GA-A, GA-H and GA-F on highly invasive human breast cancer cells. We showed that GA-A and GA-H suppressed growth (cell proliferation and colony formation) and invasive behavior (adhesion, migration and invasion) of MDA-MB-231 cells. Our results suggest that GA-A and GA-H mediate their biological effects through the inhibition of transcription factors AP-1 and NF-kappaB, resulting in the down-regulation of expression of Cdk4 and the suppression of secretion of uPA, respectively. Furthermore, the activity of ganoderic acids is linked to the hydroxylation in the position 7 and 15 (GA-A) and 3 (GA-H) in their triterpene lanostane structure. In conclusion, hydroxylated triterpenes from G. lucidum could be promising natural agents for the therapy of invasive breast cancers.
Sigma-Aldrich
Ganoderic acid A, ≥98% (HPLC)