Merck
  • Conformational preferences of natural and C3-modified epothilones in aqueous solution.
Conformational preferences of natural and C3-modified epothilones in aqueous solution.
Journal of medicinal chemistry (2008-02-15)
Máté Erdélyi, Bernhard Pfeiffer, Kurt Hauenstein, Jörg Fohrer, Jürg Gertsch, Karl-Heinz Altmann, Teresa Carlomagno
ABSTRACT
The conformational properties of the microtubule-stabilizing agent epothilone A ( 1a) and its 3-deoxy and 3-deoxy-2,3-didehydro derivatives 2 and 3 have been investigated in aqueous solution by a combination of NMR spectroscopic methods, Monte Carlo conformational searches, and NAMFIS calculations. The tubulin-bound conformation of epothilone A ( 1a), as previously proposed on the basis of solution NMR data, was found to represent a significant fraction of the ensemble of conformations present for the free ligands in aqueous solution.
Sigma-Aldrich
(−)-Epothilone A, from Sorangium cellulosum, >95% (HPLC), solid