Merck
Superhydrophobic surfaces for extreme environmental conditions.
Proceedings of the National Academy of Sciences of the United States of America (2020-10-21)
Henry Lambley, Thomas M Schutzius, Dimos Poulikakos
ABSTRACT
Superhydrophobic surfaces for repelling impacting water droplets are typically created by designing structures with capillary (antiwetting) pressures greater than those of the incoming droplet (dynamic, water hammer). Recent work has focused on the evolution of the intervening air layer between droplet and substrate during impact, a balance of air compression and drainage within the surface texture, and its role in affecting impalement under ambient conditions through local changes in the droplet curvature. However, little consideration has been given to the influence of the intervening air-layer thermodynamic state and composition, in particular when departing from standard atmospheric conditions, on the antiwetting behavior of superhydrophobic surfaces. Here, we explore the related physics and determine the working envelope for maintaining robust superhydrophobicity, in terms of the ambient pressure and water vapor content. With single-tier and multitier superhydrophobic surfaces and high-resolution dynamic imaging of the droplet meniscus and its penetration behavior into the surface texture, we expose a trend of increasing impalement severity with decreasing ambient pressure and elucidate a previously unexplored condensation-based impalement mechanism within the texture resulting from the compression, and subsequent supersaturation, of the intervening gas layer in low-pressure, humid conditions. Using fluid dynamical considerations and nucleation thermodynamics, we provide mechanistic understanding of impalement and further employ this knowledge to rationally construct multitier surfaces with robust superhydrophobicity, extending water repellency behavior well beyond typical atmospheric conditions. Such a property is expected to find multifaceted use exemplified by transportation and infrastructure applications where exceptional repellency to water and ice is desired.
Sigma-Aldrich
Trichlorovinylsilane, 97%