Merck
  • The Valuable Impacts of Halophytic Genus Suaeda; Nutritional, Chemical, and Biological Values.
The Valuable Impacts of Halophytic Genus Suaeda; Nutritional, Chemical, and Biological Values.
Medicinal chemistry (Shariqah (United Arab Emirates)) (2020-02-25)
Hamdoon A Mohammed
ABSTRACT
Suaeda is a halophytic genus belonging to the Amaranthaceae family and can survive in the high salted marsh areas of the world. Suaeda plants can biosynthesize natural substances with powerful antioxidant activity and are considered as a renewable source of energy, food, and edible oil for a larger number of populations living in the harsh environment with high salinity and drought conditions. These plants also meet folk and alternative medicines' needs. The review encompasses available scientific literature related to folk medicinal uses of Suaeda plants, their nutritional values, and chemical constituents. In addition, the biological trials applied for the Suaeda plants are also part of the review. The review covers the researches from major science literature search engines and other sites representing scientific literature, i.e., Scifinder, Google Scholar, PubMed, ScienceDirect, Scopus, and Google. The searches were programmed on the advance options available in the search engines and are latest up to November 2019. The searches were exhaustive and rechecked for accuracy. The study summarizes the uses of Suaeda plants as a remedy for various ailments due to their contents from the polyphenols and flavonoids. The comparatively large amounts of fixed oils, minerals, and vitamins in Suaeda plants have also made them potential renewable sources for foods.
Sigma-Aldrich
Quinazoline, 99%