Merck
  • Synthesis of sheet-like polypyrrole nanowires for the microextraction of trace residues of pyrethroid pesticides in human plasma and molecular dynamics-aided study of adsorption mechanism.
Synthesis of sheet-like polypyrrole nanowires for the microextraction of trace residues of pyrethroid pesticides in human plasma and molecular dynamics-aided study of adsorption mechanism.
Journal of chromatography. A (2020-10-20)
Yuge Hou, Ying Li, Wensi Huang, Makoto Tsunoda, Yingxia Zhang, Xi Xie, Rong Wang, Lushuang Li, Wenting Hu, Yanting Song, Sulan Luo
ABSTRACT
The synthesized sheet-like polypyrrole (ppy) nanowires were used as solid phase extraction materials, followed by gas chromatography-mass spectrometry (GC-MS) for the detection of traces residues of pyrethroid pesticides in human plasma. A multiresidue method was developed and verified for the determination of trace pyrethroid residues (transfluthrin, allethrin, resmethrin, fenpropathrin, etofenprox, fenvalerate) in human plasma. In this study, using the cationic surfactant cetyltrimethylammonium bromide (CTAB) as a soft template, ppy nanowires with regular morphology were prepared by oxidative polymerization. Scanning electron microscopy (SEM), transmission electron microscopy (TEM), X-ray diffraction analysis (XRD), Fourier transform infrared spectroscopy (FT-IR), thermogravimetric analysis (TGA) and other techniques were employed for characterization. Molecular dynamics analyses were used to simulate the adsorption mechanism of each pyrethroid and ppy nanowires. Based on density analysis, molecular recognition analysis, and binding energy, the van der Waals force was considered as an important driving force for the adsorption of pyrethroids and ppy nanowires. The limits of detection (LOD) of six pyrethroids were 0.008-0051 ng mL-1, and the limits of quantification (LOQ) were 0.028-0.162 ng mL-1. The relative standard deviations of ppy nanowires were 2.12-5.09%, and the recoveries of six pyrethroids ranged from 76.9 to 110.4%. The enrichment factors were within the range of 47.09-51.30. The experimental results showed that the method could be an efficient detection method for trace residue analysis of pyrethroid pesticides in complex biological samples. It would be advantageous for clinical monitoring and toxicological studies of pyrethroids.
Product Description
Supelco
Resmethrin, PESTANAL®, analytical standard, mixture of isomers