Merck
  • INX-08189, a phosphoramidate prodrug of 6-O-methyl-2'-C-methyl guanosine, is a potent inhibitor of hepatitis C virus replication with excellent pharmacokinetic and pharmacodynamic properties.
INX-08189, a phosphoramidate prodrug of 6-O-methyl-2'-C-methyl guanosine, is a potent inhibitor of hepatitis C virus replication with excellent pharmacokinetic and pharmacodynamic properties.
Antimicrobial agents and chemotherapy (2011-03-02)
John H Vernachio, Blair Bleiman, K Dawn Bryant, Stanley Chamberlain, Damound Hunley, Jeff Hutchins, Brenda Ames, Elena Gorovits, Babita Ganguly, Andrea Hall, Alexander Kolykhalov, Yule Liu, Jerry Muhammad, Nicholas Raja, C Robin Walters, Jin Wang, Karen Williams, Joseph M Patti, Geoffrey Henson, Karolina Madela, Mohamed Aljarah, Arnaud Gilles, Christopher McGuigan
ABSTRACT
INX-08189 is an aryl-phosphoramidate of 6-O-methyl-2'-C-methyl guanosine. INX-08189 was highly potent in replicon assays, with a 50% effective concentration of 10±6 nM against hepatitis C genotype 1b at 72 h. The inhibitory effect on viral replication was rapid, with a 50% effective concentration (EC50) of 35±8 nM at 24 h. An intracellular 2'-C-methyl guanosine triphosphate (2'-C-MeGTP) concentration of 2.43±0.42 pmol/10(6) cells was sufficient to achieve 90% inhibition of viral replication. In vitro resistance studies confirmed that the S282T mutation in the NS5b gene conferred an approximately 10-fold reduction in sensitivity to INX-08189. However, the complete inhibition of S282T mutant replicons still could be achieved with an EC90 of 344±170 nM. Drug combination studies of INX-08189 and ribavirin indicated significant synergy in antiviral potency both in wild-type and S282T-expressing replicons. Genotype 1b replicons could be cleared after 14 days of culture when exposed to as little as 20 nM INX-08189. No evidence of mitochondrial toxicity was observed after 14 days of INX-08189 exposure in both HepG2 and CEM human cell lines. In vivo studies of rats and cynomolgus monkeys demonstrated that 2'-C-MeGTP concentrations in liver equivalent to the EC90 could be attained after a single oral dose of INX-08189. Rat liver 2'-C-MeGTP concentrations were proportional to dose, sustained for greater than 24 h, and correlated with plasma concentrations of the nucleoside metabolite 2'-C-methyl guanosine. The characteristics displayed by INX-08189 support its continued development as a clinical candidate for the treatment of chronic HCV infection.
Sigma-Aldrich
BMS-986094, ≥98% (HPLC)
Pricing and availability is not currently available.