Merck
  • Meat quality and lipid fatty acid profile from wild thrush (Turdus philomelos), woodcock (Scolopax rusticola) and starling (Sturnus vulgaris): a preliminary comparative study.
Meat quality and lipid fatty acid profile from wild thrush (Turdus philomelos), woodcock (Scolopax rusticola) and starling (Sturnus vulgaris): a preliminary comparative study.
Lipids in health and disease (2020-06-04)
Simona Tarricone, Maria Antonietta Colonna, Carlo Cosentino, Francesco Giannico, Marco Ragni
ABSTRACT
The present study aimed to evaluate the nutritional proximate composition, some qualitative traits and fatty acid profile of meat from wild thrush, woodcock and starling hunted in Southern Italy in 2017 and 2018. Nutritive composition and physical traits of meat and lipid fatty acid profile were evaluated in breast muscle (Pectoralis major) of gamebirds. From findings, the meat pH was significantly (P < 0.001) higher in starling when compared to the other two species. Thrush meat was significantly (P = 0.002) darker and had higher redness (P < 0.001) and yellowness (P = 0.004) in comparison to starling and woodcock. Thrush breast muscle showed the highest (P < 0.001) level of lipids and lowest (P < 0.001) protein content. Meat from thrush showed the best lipid fatty acid profile based on the higher (P < 0.001) monounsaturated fatty acids (MUFA) and lower (P < 0.001) saturated fatty acids (SFA) concentrations. Starling breast muscle reported the highest (P = 0.002) polyunsaturated fatty acids (PUFA) level compared to both thrush and woodcock, whereas no differences were detected on total n-3. The ratio n-6/n-3 was higher (P = 0.001) in starling muscle. Thrush breast muscle had the lowest (P < 0.001) atherogenic and thrombogenic indices compared to the other gamebirds. The findings indicated that meat from the three investigated gamebirds species may represent a healthily lipid food source for human consumption in relation to the prevention of cardiovascular diseases.