Merck
  • A Nonlive Preparation of Chromobacterium sp. Panama (Csp_P) Is a Highly Effective Larval Mosquito Biopesticide.
A Nonlive Preparation of Chromobacterium sp. Panama (Csp_P) Is a Highly Effective Larval Mosquito Biopesticide.
Applied and environmental microbiology (2020-03-30)
Eric P Caragata, Luisa M Otero, Jenny S Carlson, Nahid Borhani Dizaji, George Dimopoulos
ABSTRACT
Given the continued high prevalence of mosquito-transmitted diseases, there is a clear need to develop novel disease and vector control strategies. Biopesticides of microbial origin represent a promising source of new approaches to target disease-transmitting mosquito populations. Here, we describe the development and characterization of a novel mosquito biopesticide, derived from an air-dried, nonlive preparation of the bacterium Chromobacterium sp. Panama (family: Neisseriaceae). This preparation rapidly and effectively kills the larvae of prominent mosquito vectors, including the dengue and Zika vector Aedes aegypti and the human malaria vector Anopheles gambiae During semi-field trials in Puerto Rico, we observed high efficacy of the biopesticide against field-derived A. aegypti populations, and against A. aegypti and Culex species larvae in natural breeding water, indicating the suitability of the biopesticide for use under more natural conditions. In addition to high efficacy, the nonlive Csp_P biopesticide has a low effective dose, a long shelf life, and high heat stability and can be incorporated into attractive larval baits, all of which are desirable characteristics for a biopesticide.IMPORTANCE We have developed a novel preparation to kill mosquitoes from an abundant soil bacterium, Chromobacterium sp. Panama. This preparation is an air-dried powder containing no live bacteria, and it can be incorporated into an attractive bait and fed directly to mosquito larvae. We demonstrate that the preparation has broad spectrum activity against the larval form of the mosquitoes responsible for the transmission of malaria and the dengue, chikungunya, yellow fever, West Nile, and Zika viruses, as well as mosquito larvae that are already resistant to commonly used mosquitocidal chemicals. Our preparation possesses many favorable traits: it kills at a low dosage, and it does not lose activity when exposed to high temperatures, all of which suggest that this preparation could eventually become an effective new tool for controlling mosquitoes and the diseases they spread.
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
LB Broth with agar (Lennox), Highly-referenced microbial growth powder medium with Agar, low salt, suitable for salt-sensitive E. coli culture.
Sigma-Aldrich
LB Broth (Lennox), Highly-referenced microbial growth powder medium, low salt, suitable for salt-sensitive E.coli culture.