Merck
  • Resveratrol promotes osteogenesis and alleviates osteoporosis by inhibiting p53.
Resveratrol promotes osteogenesis and alleviates osteoporosis by inhibiting p53.
Aging (2020-05-28)
Tao Yu, Zaiyan Wang, Xiaomeng You, Haichao Zhou, Wenbao He, Bing Li, Jiang Xia, Hui Zhu, Youguang Zhao, Guangrong Yu, Yuan Xiong, Yunfeng Yang
ABSTRACT
Although osteoporosis is one of the most common chronic age-related diseases, there is currently no gold standard for treatment. Evidence suggests resveratrol, a natural polyphenolic compound, may be helpful in the treatment of osteoporosis and other diseases. However, the molecular mechanisms underlying the anti-osteoporotic effects of resveratrol remain largely unknown. In the present study, KEGG pathway enrichment analysis of resveratrol-targeted genes identified 33 associated pathways, 12 of which were also involved in osteoporosis. In particular, the MDM2/p53 signaling pathway was identified as a potential key pathway among the shared pathways. In vitro experiments indicated that MDM2-mediated p53 degradation induced osteoblast differentiation, and resveratrol could partially reverse p53-dependent inhibition of osteogenic differentiation. These findings suggest resveratrol may alleviate osteoporosis at least in part by modulating the MDM2/p53 signaling pathway.