Merck
  • Reversible Hybrid Aqueous Li-CO2 Batteries with High Energy Density and Formic Acid Production.
Reversible Hybrid Aqueous Li-CO2 Batteries with High Energy Density and Formic Acid Production.
ChemSusChem (2020-02-11)
Rui Yang, Zhen Peng, Jiafang Xie, Yiyin Huang, Rahul Anil Borse, Xueyuan Wang, Maoxiang Wu, Yaobing Wang
ABSTRACT
Metal-CO2 batteries, an attractive technology for both energy storage and CO2 utilization, are typically classified into organic Li(Na)-CO2 batteries with a high energy density/output voltage and aqueous Zn-CO2 batteries with flexible chemical production. However, achieving both high-efficiency energy storage and flexible chemical production is still challenging. In this study, a reversible hybrid aqueous Li-CO2 battery is developed, integrating Li with an aqueous phase, which exhibits not only a high operating voltage and energy density but also highly selective formic acid production. Based on a Li plate as the anode, NaCl solution as the aqueous electrolyte, solid electrolyte Li1.5 Al0.5 Ge1.5 P3 O12 (LAGP) as a separator and Li+ transporter, and a bifunctional Pd-based electrocatalyst as the cathode, the resulting battery shows a high discharge voltage of up to 2.6 V, an outstanding energy conversion efficiency of above 80 %, and remarkable selectivity of CO2 -to-HCOOH conversion of up to 97 %. The related reaction mechanism is proposed as CO2 +2 Li+2 H+ ⇌HCOOH+2 Li+ .