Merck
  • Formation of metal-anion arrays within layered perovskite hosts. Preparation of a series of new metastable transition-metal oxyhalides, (MCl)LaNb(2)O(7) (M = Cr, Mn, Fe, Co).
Formation of metal-anion arrays within layered perovskite hosts. Preparation of a series of new metastable transition-metal oxyhalides, (MCl)LaNb(2)O(7) (M = Cr, Mn, Fe, Co).
Inorganic chemistry (2002-06-25)
Liliana Viciu, Gabriel Caruntu, Nicolas Royant, Julien Koenig, Weilie L Zhou, Thomas A Kodenkandath, John B Wiley
ABSTRACT
A new series of transition-metal oxyhalides (MCl)LaNb(2)O(7) (M = Cr, Mn, Fe, Co) have been prepared by a simple topochemical route. Layered perovskite hosts (ALaNb(2)O(7), A = Li, Na, K or Rb) were reacted with the corresponding anhydrous metal halides under mild reaction conditions (<400 degrees C). The compounds were examined by X-ray powder diffraction; the series appears to be isostructural with (CuCl)LaNb(2)O(7), and the layer spacings, with the exception of M = Co, follow the trend expected from transition-metal cationic radii. Thermal analysis with differential scanning calorimetry (DSC) shows the materials to be metastable where all four compounds decompose exothermically above 690 degrees C.
Sigma-Aldrich
Chromium(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis