Merck
Flavonoid Glycosides from the Seeds of Litchi chinensis.
Journal of agricultural and food chemistry (2011-02-04)
Xinya Xu, Haihui Xie, Jing Hao, Yueming Jiang, Xiaoyi Wei
ABSTRACT
Seven flavonoid glycosides, including one new (1) and five previously uncharacterized (3-7), were obtained from the seeds of lychee ( Litchi chinensis Sonn. cv. Heiye) by means of repetitive column chromatography and high-performance liquid chromatography (HPLC) preparation. They were identified as litchioside D (1), (-)-pinocembrin 7-O-neohesperidoside (2), (-)-pinocembrin 7-O-rutinoside (3), taxifolin 4'-O-β-d-glucopyranoside (4), kaempferol 7-O-neohesperidoside (5), tamarixetin 3-O-rutinoside (6), and phlorizin (7) on the basis of spectroscopic analysis and comparison of their data to the values reported in the literatures. Among them, compounds 1, 4, and 5 showed in vitro antitumor activity against A549, LAC, Hep-G2, and HeLa cell lines in the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) colorimetric assay.