Merck
  • Temperature requirements for thermal modulation in comprehensive two-dimensional gas chromatography.
Temperature requirements for thermal modulation in comprehensive two-dimensional gas chromatography.
Journal of separation science (2004-09-01)
Richard B Gaines, Glenn S Frysinger
ABSTRACT
Temperature requirements for trapping and release of compounds in a cryogenic gas loop-type GC x GC modulator were determined. Maximum trapping temperatures on the uncoated, deactivated modulator capillary were determined for compounds from C4 (bp -0.5 degrees C) to C40 (bp 522 degrees C). The liquid-nitrogen cooled gas flow rate was reduced from a high of 15.5 to 1.5 SLPM over the range to achieve the required trapping temperature. Excessive cold jet flow rates caused irreversible trapping and peak tailing for semi-volatile compounds above C26. Alternate cold jet coolants were investigated. An ice water-cooled jet was able to trap compounds with boiling points from C18 (bp 316 degrees C) to C40 and a room temperature air-cooled jet was able to trap compounds from C20 (bp 344 degrees C) to C40. The hot jet produced launch temperatures approximately 40 degrees C hotter than the elution temperature with heating time constants of 8 to 27 ms. Modulated compound peaks were symmetrical with half-height peak widths of 43 to 56 ms for compounds with little second column retention, and 70 to 75 ms for compounds with more second column retention. The liquid nitrogen-cooled loop modulator with gas flow programming was used to produce a GC x GC chromatogram for a crude oil that contained compounds from C7 to C47.