Merck
  • Precursors for cholesterol synthesis (7-dehydrocholesterol, 7-dehydrodesmosterol, and desmosterol): cholesterol/7-dehydrocholesterol ratio as an index of development and aging in PNS but not in CNS.
Precursors for cholesterol synthesis (7-dehydrocholesterol, 7-dehydrodesmosterol, and desmosterol): cholesterol/7-dehydrocholesterol ratio as an index of development and aging in PNS but not in CNS.
Journal of neurochemistry (1990-04-01)
J M Bourre, M Clément, D Gérard, R Legrand, J Chaudière
ABSTRACT
In rat sciatic nerve, the 7-dehydrocholesterol content decreased dramatically during the postnatal period and slowly during adulthood and aging. In contrast, the 7-dehydrodesmosterol content peaked at 14 days and was nearly undetectable after 60 days. The desmosterol content peaked at 21 days and was nearly undetectable after 1 year. The cholesterol content increased up to 21 days and remained nearly constant thereafter. In brain (in contrast to sciatic nerve), 7-dehydrodesmosterol and desmosterol contents decreased dramatically during development and slightly during adulthood and aging; the 7-dehydrocholesterol content peaked at 21 days and remained constant during aging. Only 7-dehydrocholesterol was dramatically more concentrated in PNS than in CNS. In brain, the cholesterol/7-dehydrocholesterol ratio increased during development and remained stable after 6 months. In contrast, in sciatic nerve, this ratio continuously increased during development and aging (950-fold between 5 days and 18 months). Thus, the cholesterol/7-dehydrocholesterol ratio is a useful biochemical index of development and aging in the PNS.