Merck
Pharmacology and laboratory testing of the oral Xa inhibitors.
Clinics in laboratory medicine (2014-08-30)
Meyer Michel Samama, Sadia Meddahi, Charles Marc Samama
ABSTRACT
New oral factor Xa inhibitors are intended to progressively substitute the oral vitamin K antagonists and parenteral indirect inhibitors of factor Xa in the prevention and treatment of venous and arterial thromboembolic episodes. This article focuses on the main clinical studies and on biological measurements of new oral factor Xa inhibitors, and addresses several safety issues. These newer agents do not require any routine laboratory monitoring of blood coagulation; however, biological tests have been developed in order to assess the plasma concentration of these drugs in several clinical settings. This article reviews these 4 oral direct factor Xa inhibitors.
Sigma-Aldrich
Betrixaban, ≥98% (HPLC)