Merck
  • The Effect of Diet Supplementation with Pomegranate and Bitter Melon on Lipidomic Profile of Serum and Cancerous Tissues of Rats with Mammary Tumours.
The Effect of Diet Supplementation with Pomegranate and Bitter Melon on Lipidomic Profile of Serum and Cancerous Tissues of Rats with Mammary Tumours.
Antioxidants (Basel, Switzerland) (2020-03-21)
Agnieszka Białek, Małgorzata Jelińska, Małgorzata Białek, Tomasz Lepionka, Małgorzata Czerwonka, Marian Czauderna
ABSTRACT
The aim of this study was to present overall lipid profile of organisms with ongoing neoplastic process and applied diet supplementation with pomegranate seed oil (PSO) and bitter melon extract (BME). The following were quantified in serum and cancerous tissues of rats suffering from mammary tumours: fatty acids, conjugated fatty acids and sterols, their oxidised metabolites (malondialdehyde and oxysterols) and lipoxygenase (LOX) metabolites of polyunsaturated fatty acids. The obtained results indicate that abnormalities in lipid metabolism accompany neoplastic process. These differences concern all classes of lipids and most pathways of their transformation, with the special emphasis on lipid peroxidation and LOX-mediated metabolism. Cancer process appears to be so detrimental that it may conceal positive influence of dietary modifications. The lack of anticarcinogenic properties of PSO and BME in this model may be due to their antioxidant properties or elevated levels of conjugated linoleic acids (CLA), which change CLA isomer activity from anti- to pro-tumorigenic. As CLA are the product of conjugated linolenic acids (CLnA) endogenous metabolism, high CLA levels may be explained by applied diet enrichment.