Merck
  • Integrin alpha9 beta1 is a receptor for nerve growth factor and other neurotrophins.
Integrin alpha9 beta1 is a receptor for nerve growth factor and other neurotrophins.
Journal of cell science (2008-01-31)
Izabela Staniszewska, Ilker K Sariyer, Shimon Lecht, Meghan C Brown, Erin M Walsh, George P Tuszynski, Mahmut Safak, Philip Lazarovici, Cezary Marcinkiewicz
ABSTRACT
The integrin alpha9beta1 is a multifunctional receptor that interacts with a variety of ligands including vascular cell adhesion molecule 1, tenascin C and osteopontin. We found that this integrin is a receptor for nerve growth factor (NGF) and two other neurotrophins, brain-derived neurotrophic factor and NT3, using a cell adhesion assay with the alpha9SW480 cell line. Interaction of alpha9beta1 with NGF was confirmed in an ELISA assay by direct binding to purified integrin. alpha9beta1 integrin binds to neurotrophins in a manner similar to another common neurotrophin receptor, p75(NTR) (NGFR), although alpha9beta1 activity is correlated with induction of pro-survival and pro-proliferative signaling cascades. This property of alpha9beta1 resembles the interaction of NGF with a high affinity receptor, TrkA, however, this integrin shows a low affinity for NGF. NGF induces chemotaxis of cells expressing alpha9beta1 and their proliferation. Moreover, alpha9beta1 integrin is a signaling receptor for NGF, which activates the MAPK (Erk1/2) pathway. The alpha9beta1-dependent chemotactic ability of NGF appears to result from the activation of paxillin.
Sigma-Aldrich
Anti-Integrin β1 Antibody, activated, clone B44, azide free, clone B44, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse