Merck
  • MACC1 Induces Tumor Progression in Transgenic Mice and Colorectal Cancer Patients via Increased Pluripotency Markers Nanog and Oct4.
MACC1 Induces Tumor Progression in Transgenic Mice and Colorectal Cancer Patients via Increased Pluripotency Markers Nanog and Oct4.
Clinical cancer research : an official journal of the American Association for Cancer Research (2016-01-14)
Clara Lemos, Markus S Hardt, Manisha Juneja, Cynthia Voss, Susann Förster, Boris Jerchow, Wolfram Haider, Hendrik Bläker, Ulrike Stein
ABSTRACT
We have previously identified the gene MACC1 as a strong prognostic biomarker for colorectal cancer metastasis and patient survival. Here, we report for the first time the generation of transgenic mouse models for MACC1. We generated mice with transgenic overexpression of MACC1 in the intestine driven by the villin promoter (vil-MACC1) and crossed them with Apc(Min) mice (vil-MACC1/Apc(Min)). vil-MACC1/Apc(Min) mice significantly increased the total number of tumors (P = 0.0056). This was particularly apparent in large tumors (≥3-mm diameter; P = 0.0024). A detailed histopathologic analysis of these lesions demonstrated that the tumors from the vil-MACC1/Apc(Min) mice had a more invasive phenotype and, consequently, showed a significantly reduced survival time than Apc(Min) mice (P = 0.03). Molecular analysis revealed an increased Wnt and pluripotency signaling in the tumors of vil-MACC1/Apc(Min) mice. Specifically, we observed a prominent upregulation of the pluripotency markers Oct4 and Nanog in these tumors compared with Apc(Min) controls. Finally, we could also validate that Oct4 and Nanog are regulated by MACC1 in vitro and strongly correlate with MACC1 levels in a cohort of 60 tumors of colorectal cancer patients (r = 0.7005 and r = 0.6808, respectively; P > 0.0001 and P > 0.0002, respectively). We provide proof of principle that MACC1-induced tumor progression in colorectal cancer acts, at least in part, via the newly discovered MACC1/Nanog/Oct4 axis. These findings might have important implications for the design of novel therapeutic intervention strategies to restrict tumor progression. Clin Cancer Res; 22(11); 2812-24. ©2016 AACR.
Sigma-Aldrich
Monoclonal Anti-MACC1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0856, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid