Merck
  • Polyomavirus transforms rat F111 and mouse NIH 3T3 cells by different mechanisms.
Polyomavirus transforms rat F111 and mouse NIH 3T3 cells by different mechanisms.
Journal of virology (1989-02-01)
L Raptis, J B Bolen
ABSTRACT
Polyomavirus middle tumor antigen (mT) was expressed in a line of mouse NIH 3T3 cells under control of the dexamethasone-regulatable mouse mammary tumor virus promotor. Contrary to rat F111 cells which were rendered anchorage independent by mT expression alone (L. Raptis, H. Lamfrom, and T.L. Benjamin, Mol. Cell. Biol. 5:2476-2487, 1985), mT-producing NIH 3T3 cells were unable to grow in agar even after full mT induction. The mT:pp60c-src-associated phosphatidylinositol kinase was activated in these cells to a degree similar to that in fully transformed cells expressing the small and large T antigens, in addition to mT. We therefore propose that the stimulation of this phosphatidylinositol kinase, although apparently necessary, is not sufficient for transformation of NIH 3T3 cells by polyomavirus.