- Fatty acid composition and conjugated linoleic acid content in different carcass parts of Dağlıç lambs.
Fatty acid composition and conjugated linoleic acid content in different carcass parts of Dağlıç lambs.
TheScientificWorldJournal (2014-02-14)
Ali Karabacak, İbrahim Aytekin, Saim Boztepe
PMID24523647
ABSTRACT
This study was conducted to compare fatty acid composition and content of conjugated linoleic acid (CLA) in different regions of sheep carcasses. Lambs of the Dağlıç breed were used for this purpose. Subsequent to a 68-day period of intensive fattening, fatty acids were examined in samples taken from the legs, shoulders, breasts, and ribs of lamb carcasses. According to the analysis, in leg, shoulder, breast, and rib, respectively, total saturated fatty acids (SFA) were found to be 40.38, 42.69, 42.56, and 40.27%, unsaturated fatty acids (MUFA) were found to be 40.38, 44.17, 46.17, and 49.50%, polyunsaturated fatty acids (PUFA) were found to be 4.79, 4.29, 3.80, and 3.72%, and CLAs were found to be 1.49, 1.69, 1.53, and 1.59%.