Merck
  • Liposomal mupirocin holds promise for systemic treatment of invasive Staphylococcus aureus infections.
Liposomal mupirocin holds promise for systemic treatment of invasive Staphylococcus aureus infections.
Journal of controlled release : official journal of the Controlled Release Society (2019-11-13)
Oliver Goldmann, Ahuva Cern, Mathias Müsken, Manfred Rohde, William Weiss, Yechezkel Barenholz, Eva Medina
ABSTRACT
Staphylococcus aureus is a major cause of severe invasive infections. The increasing incidence of infections caused by antibiotic-resistant strains such as methicillin-resistant S. aureus (MRSA), calls for exploration of new approaches to treat these infections. Mupirocin is an antibiotic with a unique mode of action that is active against MRSA, but its clinical use is restricted to topical administration because of its limited plasma stability and rapid degradation to inactive metabolites. Mupirocin was identified by a machine learning approach to be suitable for nano-liposome encapsulation. The computational predictions were verified experimentally and PEGylated nano-liposomal formulation of mupirocin (Nano-mupirocin) was developed. The aim of this study was to investigate the efficacy of this formulation when administered parenterally for the treatment of S. aureus invasive infections. Nano-mupirocin exhibited prolonged half-life of active antibiotic and displayed superior antimicrobial activity against S. aureus than free mupirocin in the presence of plasma. Parenteral application of Nano-mupirocin in a murine model of S. aureus bloodstream infection resulted in improved antibiotic distribution to infected organs and in a superior therapeutic efficacy than the free drug. Parenterally administered Nano-mupirocin was also more active against MRSA than free mupirocin in a neutropenic murine lung infection model. In addition, Nano-mupirocin was very efficiently taken up by S. aureus-infected macrophages via phagocytosis leading to enhanced delivery of mupirocin in the intracellular niche and to a more efficient elimination of intracellular staphylococci. The outcome of this study highlights the potential of Nano-mupirocin for the treatment of invasive MRSA infections and support the further clinical development of this effective therapeutic approach.
Product Description
Avanti
18:1 Liss Rhod PE, Avanti Research - A Croda Brand
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
100 mL
Estimated to ship on April 29, 2025
$56.90
To order products, please contact your local dealer.