- Characterization of locomotor response to psychostimulants in the parthenogenetic marbled crayfish (Procambarus fallax forma virginalis): A promising model for studying the neural and molecular mechanisms of drug addiction.
Characterization of locomotor response to psychostimulants in the parthenogenetic marbled crayfish (Procambarus fallax forma virginalis): A promising model for studying the neural and molecular mechanisms of drug addiction.
Although scientific research using mammalian models has made great strides in uncovering the enigmatic neural and molecular mechanisms orchestrating the state of drug addiction, a complete understanding has thus far eluded researchers. The complexity of the task has led to the use of invertebrate model systems to complement the research of drug-induced reward in mammalian systems. Invertebrates, such as crayfish, offer excellent model systems to help reveal the underlying mechanisms of drug addiction as they retain the ancestral neural reward circuit that is evolutionarily conserved across taxa, and they possess relatively few, large neurons, laid out in an accessible, modularly organized nervous system. Crayfish offer the benefits of delineated developmental life stages, a large body size suitable for a variety of experimental methods, and stereotyped behaviors. Unique among crayfish is the parthenogenetic marbled crayfish (Procambarus fallax forma virginalis), a species of asexually reproducing, genetically identical clones. With the benefits of reduced individual variation, high fecundity, and easy lab husbandry, the marbled crayfish would make a particularly powerful addition to the animal model repertoire. Here we characterize the locomotor response of juvenile P. f. f. virginalis exposed to the psychostimulant, d-amphetamine sulfate. Custom video-tracking software was used to record the movement patterns of juveniles exposed to water infused with varying concentrations of d-amphetamine sulfate. ANOVA demonstrated that crayfish locomotion was significantly impacted by drug concentration. These psychostimulant effects provide the foundation of P. f. f. virginalis as a model for parsing the neural and molecular mechanisms of drug addiction.