Merck
  • Poly(amine-co-ester) nanoparticles for effective Nogo-B knockdown in the liver.
Poly(amine-co-ester) nanoparticles for effective Nogo-B knockdown in the liver.
Journal of controlled release : official journal of the Controlled Release Society (2019-05-06)
Jiajia Cui, Alexandra S Piotrowski-Daspit, Junwei Zhang, Mingjie Shao, Laura G Bracaglia, Teruo Utsumi, Young-Eun Seo, Jenna DiRito, Eric Song, Christine Wu, Asuka Inada, Gregory T Tietjen, Jordan S Pober, Yasuko Iwakiri, W Mark Saltzman
ABSTRACT
Degradable poly(amine-co-ester) (PACE) terpolymers hold tremendous promise for siRNA delivery because these materials can be formulated into delivery vehicles with highly efficient siRNA encapsulation, providing effective knockdown with low toxicity. Here, we demonstrate that PACE nanoparticles (NPs) provide substantial protein knockdown in human embryonic kidney cells (HEK293) and hard-to-transfect primary human umbilical vein endothelial cells (HUVECs). After intravenous administration, NPs of solid PACE (sPACE)-synthesized with high monomer content of a hydrophobic lactone-accumulated in the liver and, to a lesser extent, in other tissues. Within the liver, a substantial fraction of sPACE NPs were phagocytosed by liver macrophages, while a smaller fraction of NPs accumulated in hepatic stellate cells and liver sinusoidal endothelial cells, suggesting that sPACE NPs could deliver siRNA to diverse cell populations within the liver. To test this hypothesis, we loaded sPACE NPs with siRNA designed to knockdown Nogo-B, a protein that has been implicated in the progression of alcoholic liver disease and liver fibrosis. These sPACE:siRNA NPs produced up to 60% Nogo-B protein suppression in the liver after systemic administration. We demonstrate that sPACE NPs can effectively deliver siRNA therapeutics to the liver to mediate protein knockdown in vivo.