Merck
  • Selective BTK inhibition improves bendamustine therapy response and normalizes immune effector functions in chronic lymphocytic leukemia.
Selective BTK inhibition improves bendamustine therapy response and normalizes immune effector functions in chronic lymphocytic leukemia.
International journal of cancer (2018-11-24)
Eriong Lee-Vergés, Bola S Hanna, Haniyeh Yazdanparast, Vanina Rodríguez, Marta Leonor Rodríguez, Ariadna Giró, Anna Vidal-Crespo, Laia Rosich, Virginia Amador, Marta Aymerich, Neus Villamor, Julio Delgado, Peter Lichter, Patricia Pérez-Galán, Mònica López-Guerra, Elías Campo, Martina Seiffert, Dolors Colomer
ABSTRACT
The Bruton's tyrosine kinase (BTK) inhibitor ibrutinib has been shown to be highly effective in patients with chronic lymphocytic leukemia (CLL) and is approved for CLL treatment. Unfortunately, resistance and intolerance to ibrutinib has been observed in several studies, opening the door for more specific BTK inhibitors. CC-292 (spebrutinib) is a BTK inhibitor with increased specificity for BTK and less inhibition of other kinases. Our in vitro studies showed that CC-292 potently inhibited B-cell receptor signaling, activation, proliferation and chemotaxis of CLL cells. In in vivo studies using the adoptive transfer TCL1 mouse model of CLL, CC-292 reduced tumor load and normalized tumor-associated expansion of T cells and monocytes, while not affecting T cell function. Importantly, the combination of CC-292 and bendamustine impaired CLL cell proliferation in vivo and enhanced the control of CLL progression. Our results demonstrate that CC-292 is a specific BTK inhibitor with promising performance in combination with bendamustine in CLL. Further clinical trials are warranted to investigate the therapeutic efficacy of this combination regimen.