Merck
  • Metabolite identification of tanshinol borneol ester in rats by high-performance liquid chromatography combined with electrospray ionization quadrupole time-of-flight mass spectrometry.
Metabolite identification of tanshinol borneol ester in rats by high-performance liquid chromatography combined with electrospray ionization quadrupole time-of-flight mass spectrometry.
Biomedical chromatography : BMC (2018-11-18)
Jie Wang, Cuicui Ma, Qiannan Li, Xing Wang, Yang Yang, Lingjian Yang, Wei Jiang, Sha Liao, Shixiang Wang, Pu Jia, Ye Zhao, Xiaohui Zheng
ABSTRACT
Tanshinol borneol ester (DBZ) is a potential drug candidate composed of danshensu and borneol. It shows anti-ischemic and anti-atherosclerosis activity. However, little is known about its metabolism in vivo. This research aimed to elucidate the metabolic profile of DBZ through analyzing its metabolites using high-performance liquid chromatography combined with electrospray ionization quadrupole time-of-flight mass spectrometry. Chromatographic separation was performed on an Agilent TC-C18 column (150 × 4.6 mm, 5.0 μm) with gradient elution using methanol and water containing 0.2% (v/v) formic acid as the mobile phase. Metabolite identification involved analyzing the retention behaviors, changes in molecular weights and MS/MS fragment patterns of DBZ and its metabolites. As a result, 20 potential metabolites were detected and tentatively identified in rat plasma, urine and feces after administration of DBZ. DBZ could be metabolized to O-methylated DBZ, DBZ-O-glucuronide, O-methylated DBZ-O-glucuronide, hydroxylated DBZ and danshensu. Danshensu, a hydrolysis product of DBZ, could further be transformed into 12 metabolites. The proposed method was confirmed to be a reliable and sensitive alternative for characterizing metabolic pathways of DBZ and providing valuable information on its druggability.