Merck
  • Lithium chloride corrects weakness and myopathology in a preclinical model of LGMD1D.
Lithium chloride corrects weakness and myopathology in a preclinical model of LGMD1D.
Neurology. Genetics (2019-05-28)
Andrew R Findlay, Rocio Bengoechea, Sara K Pittman, Tsui-Fen Chou, Heather L True, Conrad C Weihl
ABSTRACT
To understand DNAJB6's function in skeletal muscle and identify therapeutic targets for limb-girdle muscular dystrophy 1D (LGMD1D). DNAJB6 knockout (KO) myoblasts were generated with Crispr/cas9 technology, and differentially accumulated proteins were identified using stable isotope labeling, followed by quantitative mass spectrometry. Cultured KO myotubes and mouse muscle from DNAJB6b-WT or DNAJB6b-F93L mice were analyzed using histochemistry, immunohistochemistry, and immunoblot. Mouse functional strength measures included forelimb grip strength and inverted wire hang. DNAJB6 inactivation leads to the accumulation of sarcomeric proteins and hypertrophic myotubes with an enhanced fusion index. The increased fusion in DNAJB6 KO myotubes correlates with diminished glycogen synthase kinase-β (GSK3β) activity. In contrast, LGMD1D mutations in DNAJB6 enhance GSK3β activation and suppress β-catenin and NFAT3c signaling. GSK3β inhibition with lithium chloride improves muscle size and strength in an LGMD1D preclinical mouse model. Our results suggest that DNAJB6 facilitates protein quality control and negatively regulates myogenic signaling. In addition, LGMD1D-associated DNAJB6 mutations inhibit myogenic signaling through augmented GSK3β activity. GSK3β inhibition with lithium chloride may be a therapeutic option in LGMD1D.
Sigma-Aldrich
Anti-Myosin (Skeletal, Fast) antibody, Mouse monoclonal, clone MY-32, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-hnRNP-A2/B1 antibody, Mouse monoclonal, clone DP3B3, purified from hybridoma cell culture
Pricing and availability is not currently available.