- Chemical probing of RNA and RNA/protein complexes.
Chemical probing of RNA and RNA/protein complexes.
Methods in enzymology (2009-01-01)
Zhili Xu, Gloria M Culver
PMID20946769
ABSTRACT
Chemical probing is widely used as a rapid approach for assessing RNA structure, folding, and function. In this chapter, we outline procedures for handling and using chemicals commonly used to probe nucleic acids. Detailed experimental conditions and design for footprinting and modification interference are presented herein. Protocols for RNA extraction, normalization, primer extension, and data evaluation are also provided. The methods described are designed to aid in the study of large RNAs, but with slight modifications are applicable to smaller RNAs.