- Rapid Construction of a Benzo-Fused Indoxamycin Core Enabled by Site-Selective C-H Functionalizations.
Rapid Construction of a Benzo-Fused Indoxamycin Core Enabled by Site-Selective C-H Functionalizations.
Angewandte Chemie (International ed. in English) (2016-05-21)
T Aaron Bedell, Graham A B Hone, Damien Valette, Jin-Quan Yu, Huw M L Davies, Erik J Sorensen
PMID27206223
ABSTRACT
Methods for functionalizing carbon-hydrogen bonds are featured in a new synthesis of the tricyclic core architecture that characterizes the indoxamycin family of secondary metabolites. A unique collaboration between three laboratories has engendered a design for synthesis featuring two sequential C-H functionalization reactions, namely a diastereoselective dirhodium carbene insertion followed by an ester-directed oxidative Heck cyclization, to rapidly assemble the congested tricyclic core of the indoxamycins. This project exemplifies how multi-laboratory collaborations can foster conceptually novel approaches to challenging problems in chemical synthesis.
MATERIALS