Merck
  • Enhanced photon collection enables four dimensional fluorescence nanoscopy of living systems.
Enhanced photon collection enables four dimensional fluorescence nanoscopy of living systems.
Nature communications (2018-08-18)
Luciano A Masullo, Andreas Bodén, Francesca Pennacchietti, Giovanna Coceano, Michael Ratz, Ilaria Testa
ABSTRACT
The theoretically unlimited spatial resolution of fluorescence nanoscopy often comes at the expense of time, contrast and increased dose of energy for recording. Here, we developed MoNaLISA, for Molecular Nanoscale Live Imaging with Sectioning Ability, a nanoscope capable of imaging structures at a scale of 45-65 nm within the entire cell volume at low light intensities (W-kW cm-2). Our approach, based on reversibly switchable fluorescent proteins, features three distinctly modulated illumination patterns crafted and combined to gain fluorescence ON-OFF switching cycles and image contrast. By maximizing the detected photon flux, MoNaLISA enables prolonged (40-50 frames) and large (50 × 50 µm2) recordings at 0.3-1.3 Hz with enhanced optical sectioning ability. We demonstrate the general use of our approach by 4D imaging of organelles and fine structures in epithelial human cells, colonies of mouse embryonic stem cells, brain cells, and organotypic tissues.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture