Merck
  • Protein-tyrosine phosphatase SH-PTP2 (PTPN11) is localized to 12q24.1-24.3.
Protein-tyrosine phosphatase SH-PTP2 (PTPN11) is localized to 12q24.1-24.3.
Human genetics (1995-11-01)
U Dechert, A M Duncan, L Bastien, C Duff, M Adam, F R Jirik
ABSTRACT
A 2.1-kb cDNA probe encoding the human SH2-domain containing protein-tyrosine phosphatase SH-PTP2 (PTPN11) was hybridized to human metaphase chromosomes in three independent experiments. In each instance, hybridization was maximal to chromosome 12q24.1-q24.3. The presence of SH-PTP2 cDNA crosshybridizing sequences located on a number of other chromosomes suggested that SH-PTP2-related genes or pseudogenes are present in the human genome.